Learn More
Brannan™ Soda Glass Range Finder Hydrometer
Find the right hydrometer for your needs. Brannan™ Soda Glass Range Finder Hydrometers are frequently used to help select the appropriate hydrometer for subsequent accurate determinations.
Brand:  Brannan™ 10/550/0
Product Code. 12831428
  • £45.90 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Suitable for less accurate readings and are frequently used to help select the appropriate hydrometer for subsequent accurate determinations.
Specifications
Specifications
±0.02g/mL
320 mm
0.70 to 2.00 g/mL
0.01 g/mL
Steel Shot
15.6°C