Learn More
Samco Soda Lime Glass Centrifuge Tubes
Soda Lime Glass Centrifuge Tubes can withstand max. RCF 1300 x g.
Brand:  Samco G131/71
Product Code. 12883452
  • £74.55 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Description
Description
  • Conical with rim
  • Graduated
Specifications
Specifications
Clear
Soda Lime Glass
Conical with rim
Conical with rim
1300 x g
15 mL
Centrifugation Applications
112 mm
17 mm