Learn More
Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tube
Manufactured from soda lime glass and constructed with light walls. Fisherbrand™ Soda Lime Glass Light Wall Rimmed Test Tubes are ideal for general mixing and laboratory work.
£19.08 - £55.10
Specifications
Certifications/Compliance ISO 4142
Material Soda Lime Glass
Rimmed Yes
View More Specs
Products 6
Product Code Brand Diameter (Metric) Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Length (Metric) Price Quantity & Availability  
15892255
View Documents
Fisherbrand™
90101003
10 mm 75 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15832265
View Documents
Fisherbrand™
90101010
12 mm 100 mm
£36.60
Pack of 100
In Stock
to see stock.
Add to basket
15852265
View Documents
Fisherbrand™
90101015
16 mm 125 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15872265
View Documents
Fisherbrand™
90101019
16 mm 150 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15812265
View Documents
Fisherbrand™
90101006
12 mm 75 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15842275
View Documents
Fisherbrand™
90101195
24 mm 150 mm
£55.10
Pack of 50
In Stock
to see stock.
Add to basket
Description
Description
  • Withstands moderate warming
  • Light weight
  • Complies with ISO 4142
Specifications
Specifications
ISO 4142
Soda Lime Glass
Yes