Learn More
BRAND™ Soda Lime Glass Petri Dishes
Perfect for microbial or cell culture applications, these Soda Lime Glass Petri Dishes feature excellent glass quality and workmanship.
£32.85 - £79.95
Specifications
Material Soda Lime Glass
View More Specs
Products 6
Product Code Brand Diameter (Metric) Height (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Height (Metric) Price Quantity & Availability  
10624552
View Documents Promotion Details
BRAND™
455742
100 mm 15 mm
£45.60
Pack of 10
In Stock
to see stock.
Edge
Add to basket
10400873
View Documents
BRAND™
455751
150 mm 25 mm
£79.95
Pack of 10
In Stock
to see stock.
Edge
Add to basket
10019880
View Documents Promotion Details
BRAND™
455701
40 mm 12 mm
£40.40
Pack of 10
In Stock
to see stock.
Edge
Add to basket
10450293
View Documents Promotion Details
BRAND™
455717
60 mm 15 mm
£32.85
Pack of 10
In Stock
to see stock.
Edge
Add to basket
10315571
View Documents
BRAND™
455732
80 mm 15 mm
£41.00
Pack of 10
In Stock
to see stock.
Edge
Add to basket
10039890
View Documents
BRAND™
455743
100 mm 20 mm
£38.15
Pack of 10
In Stock
to see stock.
Edge
Add to basket
Description
Description
  • Dish and lid are flat inside and outside
  • Free from bubbles and streaks
  • Cut edges are fire-polished
  • Suitable for short-term chemical exposure
Specifications
Specifications
Soda Lime Glass