missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Soda Lime Glass Rimless Test Tube
Store test reagents and material. Fisherbrand™ soda lime glass rimless test tubes come in different sizes to meet your needs.
£18.97 - £42.39
Specifications
Certifications/Compliance | ISO 4142 |
---|---|
Material | Soda Lime Glass |
Quantity | 100 Pack |
Rimmed | No |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
11982188
|
Fisherbrand™
|
10 mm | 75 mm |
£18.97
Pack of 100 |
In Stock
Log in to see stock. |
||||
11922198
|
Fisherbrand™
|
18 mm | 150 mm |
£42.39
Pack of 100 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
||||
12961031
|
Fisherbrand™
|
12 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11912198
|
Fisherbrand™
|
16 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Soda Lime Glass
- Rimless
- Disposable
Specifications
ISO 4142 | |
Soda Lime Glass | |
100 Pack | |
No |