missing translation for 'onlineSavingsMsg'
Learn More

Scientific Glass Labs™ Chromatography Tank

Clear soda-lime glass construction. Includes lid.

Brand:  Scientific Glass Labs™ 1195

Product Code. 12856843

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Soda lime glass construction
  • For use with chromatography
  • Includes lid
  • Dimensions: 230 x 90 x 230mm
Specifications

Specifications

Soda Glass
Soda Lime Glass
Tank
Glass plate
230 mm
Soda Lime Glass Tank
1 Ea.
To Separate Mixtures of Substances
225 x 85 x 230 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.