missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Soda Lime Glass Threaded Bottles

Product Code. p-8010390 Shop All Brand Products
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
2500 mL
Coating:
None
ethylene-acrylate coating
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10407231

Brand: BRAND™ 704010

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Soda-lime glass bottle features ethylene-acrylate coating that envelops the glass bottle like a protection skin.

  • Round glass narrow mouth bottles
  • Space-saving square base
  • Available ethylene-acrylate-coated or uncoated
  • The coating envelops the glass bottle like a protection skin
  • Should the bottle break, the splintering hazards are reduced considerably
  • The maximum operating temperature for coated bottles is 80 °C
  • To preserve the coating, do not clean at temperatures exceeding 60 ºC
  • With PP screw cap GL45
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 2500 mL
Coating ethylene-acrylate coating
Color Amber
Material Soda Lime Glass
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.