Learn More
Scientific Glass Labs™ Soda Lime Glass Watch Glasses
Product Code. 13112440
Click to view available options
Diameter (Metric):
40 mm
50 mm
70 mm
80 mm
90 mm
100 mm
130 mm
150 mm
200 mm
Unit Size:
Pack of 10
Pack of 5
Product Code. 13112440
Brand: Scientific Glass Labs™ 170/1/90
Only null left
Add to basket
Add to basket
Manufactured from Soda lime glass. Scientific Glass Labs™ Soda Lime Glass Watch Glasses with thin, clear glass and ground edge can be used to evaporate liquids, hold solids for weighing, or for heating small substances.
Watch glasses are useful for evaporating liquids, holding solids for weighing, or for heating small substances.
  • Manufactured from Soda lime glass
  • Thin, clear glass
  • Ground edge
Specifications
Bottom Shape Flat
Material Soda Lime Glass
Shape Round
Type Watch Glass
For Use With (Application) Evaporating Liquids, Heating Small Substances, Holding Solids for Weighing
Interior Finish Ground Edge
Diameter (Metric) 90 mm