missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 1-heptanesulfonate monohydrate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Sodium 1-heptanesulfonate monohydrate is an ion pairing reagent used in the HPLC analysis of proteins and peptides. It is utilized in high performance capillary electrophoresis analysis of peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >300°C |
Color | White |
Linear Formula | CH3(CH2)6SO3Na·H2O |
Quantity | 10 g |
Beilstein | 3731762 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 220.26 (202.24 Anhydrous) |
Percent Purity | 99% |
Physical Form | Crystalline |
Chemical Name or Material | Sodium 1-heptanesulfonate monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'