missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 1-octanesulfonate monohydrate, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Sodium 1-octanesulfonate monohydrate is used as an anionic detergent. It is an ion pairing reagent used in the HPLC analysis of proteins and peptides. Further, it is used as a surface active agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | >300°C |
Quantity | 10 g |
Solubility Information | Soluble in water. |
Formula Weight | 234.29 (216.28 Anhydrous) |
Percent Purity | ≥99% |
Physical Form | Crystalline |
Chemical Name or Material | Sodium 1-octanesulfonate monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'