missing translation for 'onlineSavingsMsg'
Learn More

Sodium 3-chloro-2-hydroxypropanesulfonate hemihydrate, 98%

Product Code. 11368895
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11368895

Brand: Thermo Scientific Alfa Aesar L14018.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 143218-48-8 | C3H8ClNaO5S | 214.592 g/mol

Sodium 3-chloro-2-hydroxypropanesulfonate hemihydrate is used to prepare negatively charged probe particles to investigate effect of nanoporosity of cellulosic fibers on their streaming potential and their interactions with cationic polyelectrolytes, to prepare surface-derivatized microcrystalline cellulose (MCC) particles having either a strong positive or a strong negative zeta potential.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 3-chloro-2-hydroxypropanesulfonate hemihydrate is used to prepare negatively charged probe particles to investigate effect of nanoporosity of cellulosic fibers on their streaming potential and their interactions with cationic polyelectrolytes, to prepare surface-derivatized microcrystalline cellulose (MCC) particles having either a strong positive or a strong negative zeta potential.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143218-48-8
Molecular Formula C3H8ClNaO5S
Molecular Weight (g/mol) 214.592
MDL Number MFCD00149699
InChI Key ZPFGAXXLEFTBEU-UHFFFAOYSA-M
Synonym sodium 3-chloro-2-hydroxypropanesulphonate hemihydrate, sodium 3-chloro-2-hydroxypropanesulfonate hydrate, 3-chloro-2-hydroxy-1-propanesulfonic acid sodium salt hydrate, sodium 3-chloro-2-hydroxypropane-1-sulfonate hydrate, c3h6clnao4s.0.5h2o, sodium 3-chloro-2-hydroxypropane-1-sulfonate-water 1/1/1, 3-chloro-2-hydroxypropanesulfonic acid sodium salt hydrate
PubChem CID 23665651
IUPAC Name sodium;3-chloro-2-hydroxypropane-1-sulfonate;hydrate
SMILES C(C(CCl)O)S(=O)(=O)[O-].O.[Na+]

Specifications

Melting Point ∽256°C (decomposition)
Quantity 100 g
Beilstein 4164376
Solubility Information Soluble in water.
Formula Weight 205.60 (196.59 Anhydrous)
Percent Purity 98%
Chemical Name or Material Sodium 3-chloro-2-hydroxypropanesulfonate hemihydrate
compliance-icons
Product Identifier
  • Sodium 3-chloro-2-hydroxypropanesulphonatehemihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Sodium 3-chloro-2-hydroxypropanesulphonatehemihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.