Learn More
Sodium acetate, 1M aq. soln., pH 4.5, RNAse free, Thermo Scientific Chemicals
Description
Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. And also used in the purification and precipitation of nucleic acids, in protein crystallization, Staining of gels in protein gel electrophoresis and HPLC. Mordant dyeing; DNA microarray studies of E. coli response. Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents. As a substrate of acetokinase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. And also used in the purification and precipitation of nucleic acids, in protein crystallization, Staining of gels in protein gel electrophoresis and HPLC. Mordant dyeing; DNA microarray studies of E. coli response. Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents. As a substrate of acetokinase.
Solubility
Fully miscible.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
pH | 4.5 |
Physical Form | Liquid |
Quantity | 250 mL |
Linear Formula | NaOOCCH3 |
Solubility Information | Fully miscible. |
IUPAC Name | sodium acetate |
Formula Weight | 82.03 |
Concentration or Composition (by Analyte or Components) | 1M aq. soln. |
Odor | Odorless |
Chemical Name or Material | Sodium acetate, RNAse free |
RUO – Research Use Only