missing translation for 'onlineSavingsMsg'
Learn More
Sodium acetate, anhydrous, 99.997% (metals basis), Thermo Scientific Chemicals Product Code.: 11301618

Sodium acetate, anhydrous, 99.997% (metals basis), Thermo Scientific Chemicals

Artikelnummer. 11301618
100 g
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
25 g
100 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Product Code. 11301618

Brand: Thermo Scientific Alfa Aesar 011005.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Sodium acetate is used as buffering agent with acetic acid. It is used in many areas like cosmetics, pharmaceuticals, agriculture, bronzing and textile industry. It is employed in production of dye materials, as a polymerization catalyst, as a polymer stabilizer, as a plating agent, preparation of gel stains, preservative in food production and flavor enhancer in the nutrient industry. Sodium acetate used in the study of lithography, photography. It is used as a base in several organic reactions in research laboratories and industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 127-09-3
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00012459
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czechShow MoreShow Less
PubChem CID 517045
ChEBI CHEBI:32954
IUPAC Name sodium acetate
SMILES [Na+].CC([O-])=O

Spezifikation

Melting Point 324°C
pH 7.5 to 9
Physical Form Powder
Quantity 100 g
Beilstein 3595639
Merck Index 14,8571
Solubility Information Very soluble in water. Soluble in alcohol.
Formula Weight 82.03
Percent Purity 99.997%
Sensitivity Hygroscopic
Density 1.528
Assay (metals basis)
Chemical Name or Material Sodium acetate, anhydrous
Mehr anzeigen Weniger anzeigen

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP18L014 Certificate TypeCertificate of Analysis Date20/03/2025 Catalog Number011005.14,011005.06,011005.22
Lot NumberT28K013 Certificate TypeCertificate of Analysis Date14/08/2024 Catalog Number011005.14,011005.06,011005.22
Lot NumberT30K030 Certificate TypeCertificate of Analysis Date12/08/2024 Catalog Number011005.14,011005.06,011005.22
Lot NumberR31K025 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number011005.14,011005.06,011005.22
Lot NumberQ10K033 Certificate TypeCertificate of Analysis Date15/04/2024 Catalog Number011005.14,011005.06,011005.22

RUO – Research Use Only

Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts
Sodium acetate, anhydrous, 99.997% (metals basis), Thermo Scientific Chemicals >

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt