Learn More
Sodium acetate, anhydrous, 99.997% (metals basis), Thermo Scientific Chemicals
Description
Sodium acetate is used as buffering agent with acetic acid. It is used in many areas like cosmetics, pharmaceuticals, agriculture, bronzing and textile industry. It is employed in production of dye materials, as a polymerization catalyst, as a polymer stabilizer, as a plating agent, preparation of gel stains, preservative in food production and flavor enhancer in the nutrient industry. Sodium acetate used in the study of lithography, photography. It is used as a base in several organic reactions in research laboratories and industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 324°C |
Color | Black |
Physical Form | Powder |
Quantity | 5 g |
Assay Percent Range | 99.997% (metals basis) |
Beilstein | 3595639 |
Solubility Information | Very soluble in water. Soluble in alcohol. |
Formula Weight | 82.03 |
Odor | Nearly odorless |
Sensitivity | Hygroscopic |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P18L014 | Certificate of Analysis | 20/03/2025 | 011005.14,011005.06,011005.22 |
T28K013 | Certificate of Analysis | 14/08/2024 | 011005.14,011005.06,011005.22 |
T30K030 | Certificate of Analysis | 12/08/2024 | 011005.14,011005.06,011005.22 |
R31K025 | Certificate of Analysis | 11/06/2024 | 011005.14,011005.06,011005.22 |
Q10K033 | Certificate of Analysis | 15/04/2024 | 011005.14,011005.06,011005.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.