missing translation for 'onlineSavingsMsg'
Learn More

Sodium Acetate Anhydrous, Certified AR for Analysis, Fisher Chemical™

Catalog Number 10010500
Click to view available options
Quantity:
250 g
500 g
1 kg
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
sodium acetate
[Na+].CC([O-])=O
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
127-09-3
C2H3NaO2
82.03
MFCD00012459
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech
517045
CHEBI:32954
sodium acetate
[Na+].CC([O-])=O
Melting Point 324°C
Color White
pH 7.5 to 9.2
Physical Form Powder/Solid
Quantity 500 g
Formula Weight 82.03g/mol
Packaging Plastic powder jar
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'