missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate Anhydrous (White Crystals or Granular Powder), Fisher BioReagents
£86.75 - £92.60
Chemical Identifiers
CAS | 127-09-3 |
---|---|
Molecular Formula | C2H3NaO2 |
Molecular Weight (g/mol) | 82.03 |
MDL Number | MFCD00012459 |
InChI Key | VMHLLURERBWHNL-UHFFFAOYSA-M |
Synonym | sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech |
PubChem CID | 517045 |
ChEBI | CHEBI:32954 |
SMILES | [Na+].CC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10103243
|
Fisher BioReagents
BP333-500 |
500 g | Poly Bottle |
£86.75
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10498913
|
Fisher BioReagents
BP333-1 |
1 kg | Glass Bottle |
£92.60
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sodium Acetate is added to DNA reactions to facilitate ethanol precipitation of the DNA.Chemical Identifiers
127-09-3 | |
82.03 | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
517045 | |
[Na+].CC([O-])=O |
C2H3NaO2 | |
MFCD00012459 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
CHEBI:32954 |
Specifications
324°C | |
White | |
Powder/Solid | |
C2H3NaO2 | |
MFCD00012459 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
[Na+].CC([O-])=O | |
82.03 | |
CHEBI:32954 | |
≥99.0% | |
Sodium Acetate Anhydrous |
127-09-3 | |
7.5 to 9.2 | |
≥99.0 % | |
CH3COONa | |
15, 8709 | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
sodium acetate | |
517045 | |
82.03 | |
1% max. (at 105°C) |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Wear personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:1
Flammability:1
Instability:1
Recommended Storage : RT