missing translation for 'onlineSavingsMsg'
Learn More

Sodium acetate hydrate, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals

Product Code. 11348828
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Sodium acetate hydrate is used in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41484-91-7
Molecular Formula C2H3NaO2
Molecular Weight (g/mol) 82.03
MDL Number MFCD00071557
InChI Key VMHLLURERBWHNL-UHFFFAOYSA-M
Synonym sodium acetate hydrate, sodium acetate monohydrate, acetic acid, sodium salt, monohydrate, c2h3o2.na.h2o, naoac water, water sodium acetate, naoac h2o, naoac.h2o, acmc-20ajp8, sodium hydroxide acetic acid
PubChem CID 23681166
SMILES [Na+].CC([O-])=O

Specifications

Melting Point 58°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9985%
Solubility Information Soluble in water.(726g/L)at 0 C.
IUPAC Name sodium acetate
Formula Weight 82.03 (Anhydrous)
Odor Odorless
Appearance White crystals
Density 1.41 g/cm3 at 20°C
Chemical Name or Material Sodium acetate hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'