missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium acetate hydrate, Puratronic™, 99.9985% (metals basis), Thermo Scientific Chemicals
£51.86 - £515.57
Chemical Identifiers
CAS | 41484-91-7 |
---|---|
Molecular Formula | C2H3NaO2 |
Molecular Weight (g/mol) | 82.03 |
MDL Number | MFCD00071557 |
InChI Key | VMHLLURERBWHNL-UHFFFAOYSA-M |
Synonym | sodium acetate hydrate, sodium acetate monohydrate, acetic acid, sodium salt, monohydrate, c2h3o2.na.h2o, naoac water, water sodium acetate, naoac h2o, naoac.h2o, acmc-20ajp8, sodium hydroxide acetic acid |
PubChem CID | 23681166 |
SMILES | [Na+].CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11358828
|
Thermo Scientific Alfa Aesar
041685.06 |
5 g |
£51.86
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11348828
|
Thermo Scientific Alfa Aesar
041685.14 |
25 g |
£182.91
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368828
|
Thermo Scientific Alfa Aesar
041685.22 |
100 g |
£515.57
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium acetate hydrate is used in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
41484-91-7 | |
82.03 | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
23681166 |
C2H3NaO2 | |
MFCD00071557 | |
sodium acetate hydrate, sodium acetate monohydrate, acetic acid, sodium salt, monohydrate, c2h3o2.na.h2o, naoac water, water sodium acetate, naoac h2o, naoac.h2o, acmc-20ajp8, sodium hydroxide acetic acid | |
[Na+].CC([O-])=O |
Specifications
58°C | |
White | |
99.9985% | |
MFCD00071557 | |
Soluble in water.(726g/L)at 0 C. | |
[Na+].CC([O-])=O | |
82.03 | |
82.03 (Anhydrous) | |
White crystals | |
Sodium acetate hydrate |
41484-91-7 | |
Powder | |
C2H3NaO2 | |
sodium acetate hydrate, sodium acetate monohydrate, acetic acid, sodium salt, monohydrate, c2h3o2.na.h2o, naoac water, water sodium acetate, naoac h2o, naoac.h2o, acmc-20ajp8, sodium hydroxide acetic acid | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
sodium;acetate;hydrate | |
23681166 | |
Odorless | |
1.45 |
Safety and Handling
missing translation for 'einecsNumber' : 204-823-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only