missing translation for 'onlineSavingsMsg'
Learn More
Sodium Acetate Trihydrate, Extra Pure, SLR, Crystal, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ Product Code.: 10625312

Sodium Acetate Trihydrate, Extra Pure, SLR, Crystal, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 10625312
1 kg, Plastic powder jar
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy

Product Code. 10625312

Brand: Fisher Chemical S/2000/60

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

C2H9NaO5, CAS Number-6131-90-4, unii-4550k0sc9b, thomaegelin, plasmafusin, sodium acetate tn, sodium acetate usan:jan, tutofusin, sodium acetate usp:jan, natrium acetate-3-wasser, acetic acid, sodium salt, trihydrate, sodium acetate trihydrate, 5kg, CHEBI:32138, White, 136.08g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6131-90-4
Molecular Formula C2H9NaO5
Molecular Weight (g/mol) 136.079
MDL Number 71557
InChI Key AYRVGWHSXIMRAB-UHFFFAOYSA-M
Synonym sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn
PubChem CID 23665404
ChEBI CHEBI:32138
IUPAC Name sodium;acetate;trihydrate
SMILES CC(=O)[O-].O.O.O.[Na+]

Specifications

Melting Point 58°C
Color White
pH 8 to 9.5
Physical Form Solid
Quantity 1 kg
Formula Weight 136.08g/mol
Packaging Plastic powder jar
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Acetate Trihydrate, Extra Pure, SLR, Crystal, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.