missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Acetate Trihydrate, Extra Pure, SLR, Crystal, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Description
C2H9NaO5, CAS Number-6131-90-4, unii-4550k0sc9b, thomaegelin, plasmafusin, sodium acetate tn, sodium acetate usan:jan, tutofusin, sodium acetate usp:jan, natrium acetate-3-wasser, acetic acid, sodium salt, trihydrate, sodium acetate trihydrate, 5kg, CHEBI:32138, White, 136.08g/mol

Specifications
Specifications
Melting Point | 58°C |
Color | White |
pH | 8 to 9.5 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 136.08g/mol |
Packaging | Plastic powder jar |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Acetate Trihydrate, Extra Pure, SLR, Crystal, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction