Learn More
Sodium acetylide, 18 wt.% slurry in xylene, AcroSeal™, Thermo Scientific Chemicals
£40.80 - £244.40
Identifiants chimiques
CAS | 1066-26-8, 1330-20-7 |
---|---|
Molecular Formula | C2HNa |
Molecular Weight (g/mol) | 48.02 |
MDL Number | MFCD00008568 |
InChI Key | SFDZETWZUCDYMD-UHFFFAOYSA-N |
Synonym | sodium acetylide, sodium ethynide, ethynyl sodium, sodium acetylide suspension, sodium acetylide na c2h, sodium ethyne, monosodium acetylide, slbnqjagykqcki-uhfffaoysa-n, sodium acetylide suspension, 18 wt. % slurry in xylene: light mineral oil purity of contained acetylide |
PubChem CID | 2733336 |
ChEBI | CHEBI:55387 |
IUPAC Name | sodium;ethyne |
SMILES | C#[C-].[Na+] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
1066-26-8, 1330-20-7 | |
48.02 | |
SFDZETWZUCDYMD-UHFFFAOYSA-N | |
2733336 | |
sodium;ethyne |
C2HNa | |
MFCD00008568 | |
sodium acetylide, sodium ethynide, ethynyl sodium, sodium acetylide suspension, sodium acetylide na c2h, sodium ethyne, monosodium acetylide, slbnqjagykqcki-uhfffaoysa-n, sodium acetylide suspension, 18 wt. % slurry in xylene: light mineral oil purity of contained acetylide | |
CHEBI:55387 | |
C#[C-].[Na+] |
Spécification
1066-26-8, 1330-20-7 | |
Low-melting Solid or Crystalline Powder | |
HC≡CNa | |
25°C | |
Solubility in water: reacts | |
C#[C-].[Na+] | |
48.02 | |
CHEBI:55387 | |
17.0 to 22.0 wt% (Total base) | |
0.9360g/mL |
White to Yellow | |
C2HNa | |
MFCD00008568 | |
sodium acetylide, sodium ethynide, ethynyl sodium, sodium acetylide suspension, sodium acetylide na c2h, sodium ethyne, monosodium acetylide, slbnqjagykqcki-uhfffaoysa-n, sodium acetylide suspension, 18 wt. % slurry in xylene: light mineral oil purity of contained acetylide | |
SFDZETWZUCDYMD-UHFFFAOYSA-N | |
sodium;ethyne | |
2733336 | |
48.02 | |
0.936 | |
Sodium acetylide |
Sécurité et traitement
GHS H Statement
Flammable liquid and vapour.
Harmful if inhaled.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
In contact with water releases flammable gas.
May be fatal if swallowed and enter
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Brush off loose particles from skin.
Immerse
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 213-908-9
- Sodium acetylide, 18 wt% slurry in xylene
Mention d'avertissement
- Danger
Catégorie de danger
- Acute toxicity Category 4
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Mention de danger
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H373-May cause damage to organs.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H226-Flammable liquid and vapour.
- EUH014-Reacts violently with water.
Conseils de prudence
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P331-Do NOT induce vomiting.
RUO – Research Use Only