missing translation for 'onlineSavingsMsg'
Learn More

Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade, Thermo Scientific Chemicals

Product Code. 11343888
Click to view available options
Quantity:
100 g
500 g
2.5 kg

Used in the laboratory as an essential ingredient of the microcosmic salt bead test for identification of metallic radicals. Also used as intermediates. Finds application in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-13-3
Molecular Formula H13NNaO8P
Molecular Weight (g/mol) 209.067
MDL Number MFCD00066970
InChI Key IQTQISLCLWJRPM-UHFFFAOYSA-M
Synonym unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate
PubChem CID 56924540
IUPAC Name azanium;sodium;hydrogen phosphate;tetrahydrate
SMILES [NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+]

Specifications

Melting Point 79°C (decomposition)
Color White
Physical Form Crystals
Quantity 2.5 kg
Solubility Information Soluble in water. Insoluble in alcohol
Formula Weight 209.07 (137.02 Anhydrous)
Grade Reagent
Odor Odorless
Appearance White crystals
Density 1.544 g/cm3 at 20°C
Chemical Name or Material Sodium ammonium hydrogen phosphate tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.