Learn More
Sodium ammonium hydrogen phosphate tetrahydrate, Reagent Grade, Thermo Scientific Chemicals
£28.54 - £271.35
Chemical Identifiers
CAS | 7783-13-3 |
---|---|
Molecular Formula | H13NNaO8P |
Molecular Weight (g/mol) | 209.067 |
MDL Number | MFCD00066970 |
InChI Key | IQTQISLCLWJRPM-UHFFFAOYSA-M |
Synonym | unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate |
PubChem CID | 56924540 |
IUPAC Name | azanium;sodium;hydrogen phosphate;tetrahydrate |
SMILES | [NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11323888
|
Thermo Scientific Alfa Aesar
040320.22 |
100 g |
£28.54
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11333888
|
Thermo Scientific Alfa Aesar
040320.36 |
500 g |
£66.13
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11343888
|
Thermo Scientific Alfa Aesar
040320.A4 |
2.5 kg |
£271.35
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used in the laboratory as an essential ingredient of the microcosmic salt bead test for identification of metallic radicals. Also used as intermediates. Finds application in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-13-3 | |
209.067 | |
IQTQISLCLWJRPM-UHFFFAOYSA-M | |
56924540 | |
[NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+] |
H13NNaO8P | |
MFCD00066970 | |
unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate | |
azanium;sodium;hydrogen phosphate;tetrahydrate |
Specifications
79°C (decomposition) | |
White | |
H13NNaO8P | |
unii-k78u8wlv4h, ammonium sodium phosphate tetrahydrate, k78u8wlv4h, phosphorsalz, acmc-20ajp4, ammonium sodium phosphate tetrahydrate mi, sodium monoammonium monohydrogen phosphate tetrahydrate, phosphoric acid, monoammonium monosodium salt, tetrahydrate, salt of phosphorus microcosmic salt, sodium ammonium phosphate tetrahydrate | |
IQTQISLCLWJRPM-UHFFFAOYSA-M | |
azanium;sodium;hydrogen phosphate;tetrahydrate | |
56924540 | |
Reagent | |
White crystals | |
Sodium ammonium hydrogen phosphate tetrahydrate |
7783-13-3 | |
Crystals | |
MFCD00066970 | |
Soluble in water. Insoluble in alcohol | |
[NH4+].O.O.O.O.OP(=O)([O-])[O-].[Na+] | |
209.067 | |
209.07 (137.02 Anhydrous) | |
Odorless | |
1.544 |
Safety and Handling
H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only