Learn More
Sodium Arsenite 0.1 M According to European Pharmacopoeia (EP), Reagecon
Fully factorized, high-purity, stable product developed and tested for titrations.
Brand: Reagecon 3005800
Code : A0
Additional Details : CAS Number : 7647-01-0 Weight : 1.00000kg Transport : UN number : 1686 Chem class : 6.1 Pack group : III
Description
- European Pharmacopoeia (EP) Reference Standard, European Pharmacopoeia (EP) Reference Material
- Accurate, stable, reproducible and fully factorized.
- Titrations performed with this product, exhibit defined stoichiometry, are quantitative, establish an equilibrium that is definitive and fast and provide unambiguous results.
- Stability is proven (either opened or unopened) for the shelf life of this product.
- Certified to a specification of ± 0.2%.
- Certified by Reagecon’s ISO 17025 Accredited Test Method (INAB Ref: 264T)
- Product is a reagent of known concentration, that is dispensed from a burette or other dispensing apparatus to a sample (analyte), until a reaction between the two liquids is judged to be complete.
- May be used for manual titrations, or automatic titrations, to determine the end points by detection as detailed in several EP monographs.
- NIST traceable.
Specifications
Accuracy ± 0.2% | |
HDPE Bottle GL45 | |
Titration and EP Monographs |
0.1 M | |
1 L | |
Liquid |
Safety and Handling
- 0.1M sodium arsenite
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 1A
Hazard Statement
- H302-Harmful if swallowed.
- H350-May cause cancer.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid, sodium hydroxide, diarsenic trioxide