missing translation for 'onlineSavingsMsg'
Learn More

Sodium Arsenite, 98+%, Extra Pure, SLR, Fisher Chemical

Brand:  Fisher Chemical S/2330/48

Code : 21

Additional Details : CAS Number : 7784-46-5 Weight : 0.10000kg Transport : UN number : 2027 Chem class : 6.1 Pack group : II


Product Code. 12897692

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

100g, Plastic powder jar

Specifications

Specifications

615°C
7784-46-5
Solid
AsNaO2
sodium arsenite, sodium metaarsenite, sodium dioxoarsenate, prodalumnol, sodanit, penite, kill-all, prodalumnol double, rat death liquid, chem pels c
[O-][As]=O.[Na+]
129.909
CHEBI:29678
Plastic powder jar
White
12
100 g
3472
PTLRDCMBXHILCL-UHFFFAOYSA-M
sodium;oxoarsinite
443495
129.91g/mol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Sodium arsenite

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 2
  • Carcinogenicity Category 1A
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H331-Toxic if inhaled.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H350-May cause cancer.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P313-Get medical advice/attention.

SDS Documents

Documents

Certificates
Special Offers

Special Offers