missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium azide, 99%, extra pure
Tienda Thermo Scientific Chemicals Productos
missing translation for 'orderingAttributeHoverText'
Quantity:
5 g
100 g
500 g
2.5 kg
missing translation for 'unitSize'
100g
2.5kg
500g
5g
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, For Azidation/Diazo transfer
Especificaciones
Especificaciones
| Boiling Point | 300.0°C |
| Melting Point | 275.0°C |
| Color | White |
| pH | 9 (5% min. soln.) |
| Physical Form | Crystalline Powder |
| Quantity | 500 g |
| Assay Percent Range | 98.5% min |
| Molecular Formula | N3Na |
| Linear Formula | NaN3 |
| Fieser | 01,1041; 02,376; 03,259; 04,440; 05,593; 15,289; 17,68 |
| Mostrar más |
Seguridad y manipulación
Nombre del producto
- Sodium azide
- Danger
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H300-Fatal if swallowed.
- EUH032-Contact with acids liberates very toxic gas.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido