Learn More
Sodium benzyloxide, 1M solution in benzyl alcohol, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 433551000
199.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
203°C to 205°C | |
20194-18-7, 100-51-6 | |
Viscous Liquid | |
C7H7NaO | |
90°C | |
LFQULJPVXNYWAG-UHFFFAOYSA-N | |
sodium;phenylmethanolate | |
23676753 | |
0.95 to 1.10M | |
1.077 |
Sodium benzyloxide | |
Colorless to Yellow | |
100 mL | |
C6H5CH2ONa | |
sodium phenylmethanolate, sodium benzyloxide, benzyloxysodium, sodium benzylate, naobn, sodium phenylmethoxide, sodium benzyloxide solution | |
C1=CC=C(C=C1)C[O-].[Na+] | |
130.12 | |
130.12 | |
AcroSeal™ Glass Bottle | |
1.0770g/mL |
Safety and Handling
- Sodium benzyloxide, 1M solution in benzyl alchol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only