missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium bicarbonate, 1M buffer soln., pH 10.0

Product Code. 15405299
Click to view available options
Quantity:
250 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

The bicarbonate buffering system is important in many different cellular processes. It is also used in the formation of acid in the lumen on the stomach. Used to neutralize the pH of the chyme leaving the stomach and entering the small intestine. It is also used to maintain pH in serum samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The bicarbonate buffering system is important in many different cellular processes. It is also used in the formation of acid in the lumen on the stomach. Used to neutralize the pH of the chyme leaving the stomach and entering the small intestine. It is also used to maintain pH in serum samples.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Concentration 1 M
pH 10.0
Physical Form Liquid
Quantity 250 mL
Solubility Information It is soluble in water.
Chemical Name or Material Sodium bicarbonate buffer soln.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'