Learn More
Thermo Scientific Chemicals Sodium bicarbonate, 1M buffer soln., pH 9.4
£57.99 - £87.44
Chemical Identifiers
CAS | 144-55-8 |
---|---|
Molecular Formula | CHNaO3 |
Molecular Weight (g/mol) | 84.01 |
MDL Number | MFCD00003528 |
InChI Key | UIIMBOGNXHQVGW-UHFFFAOYSA-M |
PubChem CID | 516892 |
ChEBI | CHEBI:32139 |
IUPAC Name | sodium;hydrogen carbonate |
SMILES | [Na+].OC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15455109
|
Thermo Scientific Chemicals
J62808.AK |
250 mL |
£57.99
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15465109
|
Thermo Scientific Chemicals
J62808.AP |
500 mL |
£87.44
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The bicarbonate buffering system is important in many different cellular processes. It is also used in the formation of acid in the lumen on the stomach. Used to neutralize the pH of the chyme leaving the stomach and entering the small intestine. It is also used to maintain pH in serum samples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThe bicarbonate buffering system is important in many different cellular processes. It is also used in the formation of acid in the lumen on the stomach. Used to neutralize the pH of the chyme leaving the stomach and entering the small intestine. It is also used to maintain pH in serum samples.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents and strong acids.
Chemical Identifiers
144-55-8 | |
84.01 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
CHEBI:32139 | |
[Na+].OC([O-])=O |
CHNaO3 | |
MFCD00003528 | |
516892 | |
sodium;hydrogen carbonate |
Specifications
144-55-8 | |
9.4 | |
CHNaO3 | |
It is soluble in water. | |
[Na+].OC([O-])=O | |
84.01 | |
CHEBI:32139 |
1 M | |
Liquid | |
MFCD00003528 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
sodium;hydrogen carbonate | |
516892 | |
Sodium bicarbonate buffer soln. |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.