missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bicarbonate 7.5% solution
Sodium Bicarbonate 7.5% solution
£7.61 - £143.00
Specifications
Content And Storage | 15 to 30°C |
---|---|
Concentration | 7.5 % |
Form | Liquid |
Product Type | Sodium Bicarbonate |
For Use With (Application) | Mammalian Cell Culture |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10308462
|
N/A
25080060 |
100 mL |
£7.61
100mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11550626
|
N/A
25080102 |
20 x 100 mL |
£143.00
Pack of 20 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sodium Bicarbonate is buffer commonly used for maintaining the pH of cell culture medium in the presence of 4–10% carbon dioxide. In addition to buffering, sodium bicarbonate provides some nutritional benefit, while rarely displaying any cell toxicity. Formulated as 75 g per liter of water, Gibco™ Sodium Bicarbonate, 7.5% Solution is perfect for supplementing dry powder medium during reconstitution. The final Sodium Bicarbonate concentration used in the medium depends on the media formulation and the carbon dioxide concentration used in the incubator.Product Use
For Research Use Only. Not for use in diagnostic procedures.
Dual-site cGMP Manufacturing and Quality System
Gibco™ Sodium Bicarbonate Solution is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard. For supply chain continuity, we offer a comparable Gibco™ Sodium Bicarbonate Solution product made in our Grand Island facility (25080-094). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standards.
Specifications
15 to 30°C | |
Liquid | |
Mammalian Cell Culture |
7.5 % | |
Sodium Bicarbonate | |
For research use only |
For Research Use Only. Not for use in diagnostic procedures.