missing translation for 'onlineSavingsMsg'
Learn More
Sodium Bicarbonate (Crystalline Powder/USP/FCC/EP/BP/JP), Fisher Chemical™ Product Code.: 12668223

Sodium Bicarbonate (Crystalline Powder/USP/FCC/EP/BP/JP), Fisher Chemical™

Product Code. 12668223
12 kg, Poly Pail
Click to view available options
Quantity:
12 kg
Packaging:
Poly Pail
This item is not returnable. View return policy

Product Code. 12668223

Brand: Fisher Chemical USS63712

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neutShow MoreShow Less
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Melting Point 270°C
Color White
pH 8.3
Physical Form Powder/Solid
Quantity 12 kg
Solubility Information Pass Test
Formula Weight 84.01
Percent Purity 99.0 to 101.0% (EP)
Grade BP/EP/FCC/JP/USP
Identification Pass Test (BP), Pass Test (EP), Pass Test (FCC), Pass Test (JP), Pass Test (USP), Pass Test (USP/FCC/EP/BP/JP)
Loss on Drying 0.25% max. (FCC), 0.25% max. (USP), 0.25% max. (USP, FCC)
Packaging Poly Pail
Decomposition Information >50°C
Chemical Name or Material Sodium Bicarbonate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium Bicarbonate (Crystalline Powder/USP/FCC/EP/BP/JP), Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.