missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Bicarbonate (Fine White Powder), Fisher BioReagents™
30.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
Description
Sodium Bicarbonate (Fine White Powder)
This sodium salt is used in buffers for molecular biology and cell culture applications.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 270°C |
Color | White |
pH | 8.3 |
Physical Form | Powder/Solid |
Quantity | 500 g |
Formula Weight | 84.01 |
Percent Purity | 99.7 to 100.3% |
Identification | Pass Test |
Packaging | Poly Bottle |
Chemical Name or Material | Sodium Bicarbonate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'