missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bicarbonate, Honeywell Fluka™
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 25 g |
Linear Formula | NaHCO3 |
UN Number | NONH for all modes of transport |
Beilstein | 4153970 |
IUPAC Name | sodium;hydrogen carbonate |
Formula Weight | 84.01g/mol |
Percent Purity | 99.7 to 100.3% |
Chemical Name or Material | Sodium bicarbonate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'