missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Sodium Bicarbonate, Powder
Part of buffering system commonly used to maintain physiological pH 7.2 to 7.4 in culture environment
Specifications
Content And Storage | 15° to 30°C |
---|---|
Form | Powder |
Product Type | Sodium Bicarbonate |
Sterility | Sterile-filtered |
Format | Plastic Bottle |
Description
Cellgro incorporates many principles developed by the pharmaceutical industry including the use of WFI quality water, raw material testing, in-process monitoring, and finished product testing.
- Molecular weight: 84.01
Specifications
15° to 30°C | |
Sodium Bicarbonate | |
Plastic Bottle |
Powder | |
Sterile-filtered | |
6.8 to 7.4 |