missing translation for 'onlineSavingsMsg'
Learn More
Sodium bicarbonate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E500, 99.0-100.5%, powder, Honeywell Fluka™ Product Code.: 15671310

Sodium bicarbonate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E500, 99.0-100.5%, powder, Honeywell Fluka™
BIOPHARMA

Product Code. 15671310 Shop All Honeywell Fluka Products
5 kg
Click to view available options
Quantity:
1 kg
5 kg
4 x 5 kg
6 x 1 kg
This item is not returnable. View return policy

Product Code. 15671310

Brand: Honeywell-Fluka 134335KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E500, 99.0-100.5%, powder

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neutShow MoreShow Less
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium;hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

Physical Form Powder
Quantity 5 kg
Linear Formula NaHCO3
UN Number NONH for all modes of transport
Beilstein 4153970
Formula Weight 84.01g/mol
Percent Purity 99 to 100.5%
Grade puriss.
Chemical Name or Material Sodium Bicarbonate, Meets analytical specification of Ph. Eur.,BP, USP, FCC

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0450866 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number13433-5KG,13433-6X1KG,13433-1KG,134331000,134330050,134330250,13433-25KG
Lot NumberA0399094 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number13433-5KG,13433-6X1KG,13433-1KG,134331000,134330050,134330250,13433-25KG
Lot NumberA0414331 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog Number13433-5KG,13433-6X1KG,13433-1KG,134331000,134330050,134330250,13433-25KG
Lot NumberA0432307 Certificate TypeCertificate of Analysis Date05/08/2021 Catalog Number13433-5KG,13433-6X1KG,13433-1KG,134331000,134330050,134330250,13433-25KG
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sodium bicarbonate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E500, 99.0-100.5%, powder, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.