missing translation for 'onlineSavingsMsg'
Learn More

Sodium bis(trimethylsilyl)amide, 98%

Product Code. 11317748
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
This item is not returnable. View return policy

Product Code. 11317748

Brand: Thermo Scientific Alfa Aesar 033505.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1070-89-9 | C6H18NNaSi2 | 183.377 g/mol

Sodium bis(trimethylsilyl)amide is used as a base in organic synthesis as well as to deprotonate other compounds containing weakly acidic O-H, S-H, and N-H bonds. It is used to prepare amines by reacting with alkyl halides. It finds application in certain organic reactions viz. generation of carbenes by dehydrohalogenation of halocarbons and Wittig reagents by deprotonation of phosphonium salts. It accelerates the polymerization of pheneylacetylene in conjunction with rhodium(I) catalysis. Furthermore, it is used for the preparation of lanthanide complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070-89-9
Molecular Formula C6H18NNaSi2
Molecular Weight (g/mol) 183.377
MDL Number MFCD00009835
InChI Key WRIKHQLVHPKCJU-UHFFFAOYSA-N
Synonym sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt
PubChem CID 2724254
IUPAC Name sodium;bis(trimethylsilyl)azanide
SMILES C[Si](C)(C)[N-][Si](C)(C)C.[Na+]

Specifications

Melting Point 171°C to 175°C
Color White
Odor Odorless
Linear Formula (CH3)3SiNNaSi(CH3)3
Quantity 50 g
UN Number UN3096
Sensitivity Moisture sensitive
Solubility Information Soluble in hexane,toluene,ether,terahydrofuran,benzene and toluene.
Formula Weight 183.38
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Sodium bis(trimethylsilyl)amide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.