missing translation for 'onlineSavingsMsg'
Learn More

Sodium bis(trimethylsilyl)amide, pure, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals

£69.85 - £266.33

Specifications

Chemical Name or Material Sodium bis(trimethylsilyl)amide
Color Orange to Brown
Name Note 2M Solution in THF
CAS 1070-89-9, 109-99-9
Molecular Formula C6H18NNaSi2
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10053530
View Documents Promotion Details
Thermo Scientific Acros
277851000
100 mL AcroSeal™ Glass bottle
£69.85
100mL
Please to purchase this item. Need a web account? Register with us today!
10379150
View Documents
Thermo Scientific Acros
277858000
800 mL AcroSeal™ Glass Bottle
£266.33
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium bis(trimethylsilyl)amide
2M Solution in THF
C6H18NNaSi2
MFCD00009835
0.9160g/mL
WRIKHQLVHPKCJU-UHFFFAOYSA-N
sodium;bis(trimethylsilyl)azanide
2724254
38 to 42%
0.916
Liquid
Orange to Brown
1070-89-9, 109-99-9
[(CH3)3Si]2NNa
01,1046; 03,261; 04,442; 06,529; 12,441; 16,307
sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt
C[Si](C)(C)[N-][Si](C)(C)C.[Na+]
183.377
183.38
Pure
−21°C
Videos
Safety and Handling

Safety and Handling

GHS P Statement Causes severe skin burns and eye damage. May cause respiratory irritation. Highly flammable liquid and vapor. Suspected of causing cancer. Suspected of causing genetic defects. Harmful to aquatic life with long lasting effects. Reacts violently with water. May form explosive peroxides.

GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Immediately call a POISON CENTER or doctor/physician. Avoid breathing dust/fume/gas/mist/vapors/spray. Store in a dry place. Store in a closed container. Keep container tightly closed.

Danger

missing translation for 'einecsNumber' : 213-983-8

missing translation for 'tsca' : TSCA

Product Identifier
  • Sodium bis(trimethylsilyl)amide, 2M solution in THF

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only