Learn More
Sodium bis(trimethylsilyl)amide, pure, 2M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 277851000
56.65 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 29
Additional Details : CAS Number : 1070-89-9 Weight : 0.09160kg Transport : UN number : 2924 Chem class : 3 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium bis(trimethylsilyl)amide | |
2M Solution in THF | |
C6H18NNaSi2 | |
MFCD00009835 | |
0.9160g/mL | |
WRIKHQLVHPKCJU-UHFFFAOYSA-N | |
sodium;bis(trimethylsilyl)azanide | |
2724254 | |
38 to 42% | |
AcroSeal™ Glass bottle | |
100 mL | |
Liquid |
Orange to Brown | |
1070-89-9,109-99-9 | |
[(CH3)3Si]2NNa | |
01,1046; 03,261; 04,442; 06,529; 12,441; 16,307 | |
sodium bis trimethylsilyl amide, n-sodiohexamethyldisilazane, sodium hexamethyldisilazide, nahmds, sodiobis trimethylsilyl amine, sodium bis trimethylsilyl azanide, n-sodium hexamethyldisilazane, sodium-bis trimethylsilyl amide, hexamethyldisilazane sodium salt | |
C[Si](C)(C)[N-][Si](C)(C)C.[Na+] | |
183.377 | |
183.38 | |
Pure | |
0.916 | |
−21°C |
Safety and Handling
- Sodium bis(trimethylsilyl)amide, 2M solution in THF
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only