missing translation for 'onlineSavingsMsg'
Learn More

Sodium bisulfate, 92%, technical

Product Code. 10164600 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
1kg
2.5kg
25g
This item is not returnable. View return policy

Product Code. 10164600

Brand: Thermo Scientific Acros 214850250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7681-38-1 | HNaO4S | 120.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-38-1
Molecular Formula HNaO4S
Molecular Weight (g/mol) 120.05
MDL Number MFCD00003531
InChI Key WBHQBSYUUJJSRZ-UHFFFAOYSA-M
Synonym sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect
PubChem CID 516919
IUPAC Name sodium hydrogen sulfate
SMILES [Na+].OS([O-])(=O)=O

Specifications

Melting Point 177.0°C to 180.0°C
Color White to Yellow
Physical Form Granules
Quantity 25 g
Assay Percent Range 92%
Linear Formula NaHSO4
Merck Index 15, 8722
Solubility Information Solubility in water: soluble. Other solubilities: decomposes by alcohol into na2so4 and h2so4
Formula Weight 120.06
Percent Purity 92%
Grade Technical
Packaging Glass bottle
Specific Gravity 2.1
Density 2.1000g/mL
Chemical Name or Material Sodium bisulfate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium bisulfate, tech., granular
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.