missing translation for 'onlineSavingsMsg'
Learn More

Sodium bisulfite solution, Purum, ∼40%, Honeywell Fluka™

Product Code. 15611320
Click to view available options
Quantity:
1 L
2.5 L
25 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Purum, ∼40%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7631-90-5, 7732-18-5
Molecular Formula NaO3S
Molecular Weight (g/mol) 103.05
MDL Number MFCD00003530
InChI Key DWAQJAXMDSEUJJ-UHFFFAOYSA-L
Synonym sodium bisulfite, sodium hydrogen sulfite, sodium bisulphite, sodium hydrogensulfite, sodium sulfhydrate, monosodium sulfite, uantax sbs, sulfurous acid, monosodium salt, caswell no. 750, hydrogen sodium sulfite
PubChem CID 23665763
ChEBI CHEBI:26709
SMILES [Na+].[O-]S([O-])=O

Specifications

Boiling Point 100°C
CAS Min % ≥50.0000%, ≥25.0000%
CAS Max % <75.0000%, <50.0000%
Quantity 25 L
Linear Formula CH3CH2OH
UN Number 3082
IUPAC Name sodium sulfite
Formula Weight 104.06
Percent Purity 4%
Grade purum
Concentration or Composition (by Analyte or Components) 40%
Packaging Poly Pail
Density 1.31 - 1.35 g/cm3
Chemical Name or Material Sodium bisulfite solution, Purum, ∼40%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'