missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bisulfite solution, Purum, ∼40%, Honeywell Fluka™
Click to view available options
Quantity:
1 L
2.5 L
25 L
Description
- ≥95% for general laboratory use.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 L |
Linear Formula | NaHSO3 |
UN Number | 3082 |
IUPAC Name | sodium;hydrogen sulfite |
Formula Weight | 104.06g/mol |
Grade | purum |
Concentration or Composition (by Analyte or Components) | 40% |
Chemical Name or Material | Sodium Bisulfite Solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'