missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sodium borate, 0.5M buffer soln., pH 8.5

Product Code. p-7042546
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
This item is not returnable. View return policy

Product Code. 15405199

Brand: Thermo Scientific Chemicals J62902.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1330-43-4 | B4Na2O7 | 201.21 g/mol

This product is ideally suited for many biological applications, including biochemical, molecular biological, cell biological, histological, and immunological research and lab procedures. It can be used to dilute substances and in coating procedures. Additives such as Tween 20 and milk powder can be used to add function as washing buffer or blocking buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This product is ideally suited for many biological applications, including biochemical, molecular biological, cell biological, histological, and immunological research and lab procedures. It can be used to dilute substances and in coating procedures. Additives such as Tween 20 and milk powder can be used to add function as washing buffer or blocking buffer.

Solubility
It is soluble in water.

Notes
Store at 25 degrees C. This product is relatively stable under this condition. Best result obtained with use within 6-8 months of purchase.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-43-4
Molecular Formula B4Na2O7
Molecular Weight (g/mol) 201.21
MDL Number MFCD00081185
InChI Key NXSVMNYRVBQCMO-UHFFFAOYSA-N
IUPAC Name disodium bicyclo[3.3.1]tetraboroxane-3,7-bis(ylium)-1,5-diuide-1,5-bis(olate)
SMILES [Na+].[Na+].[O-][B-]12O[B+]O[B-]([O-])(O[B+]O1)O2

Specifications

Concentration 0.5 M
pH 8.5
Physical Form Liquid
Quantity 250 mL
Solubility Information It is soluble in water.
Chemical Name or Material Sodium borate buffer soln.
compliance-icons compliance-icons
Product Identifier
  • Sodium borate, 0.5M buffer solution, pH 8.5
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Disodium tetraborate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.