missing translation for 'onlineSavingsMsg'
Learn More

Sodium Borate, 10-Hydrate, N.F., J.T. Baker™

Product Code. 15538144
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1303-96-4
Molecular Formula Na2B4O7·10H2O

Specifications

Quantity 500 g
Formula Weight 381.37
Grade NF
Packaging Poly Bottle
Specific Gravity 1L = 1.73kg
Chemical Name or Material Sodium Borate, 10-Hydrate
compliance-icons
Product Identifier
  • SODIUM BORATE,10-HYDR
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : di-Sodium tetraborate decahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.