Learn More
Sodium borodeuteride, 98 atom% D, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 15681-89-7 |
---|---|
Molecular Formula | BH4Na |
Molecular Weight (g/mol) | 37.83 |
MDL Number | MFCD00003519 |
InChI Key | YOQDYZUWIQVZSF-UHFFFAOYSA-N |
Synonym | sodium borodeuteride, sodium tetradeuterioboron, sodium borohydride-d4, sodium 2h4 tetrahydroborate 1-, sulfatase, borate 1-, tetrahydro-d4-, sodium, nabd4, sodium 2h4 boranuide, deuterated sodium borohydride |
PubChem CID | 23673181 |
SMILES | [BH4-].[Na+] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMRChemical Identifiers
15681-89-7 | |
37.83 | |
YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
23673181 |
BH4Na | |
MFCD00003519 | |
sodium borodeuteride, sodium tetradeuterioboron, sodium borohydride-d4, sodium 2h4 tetrahydroborate 1-, sulfatase, borate 1-, tetrahydro-d4-, sodium, nabd4, sodium 2h4 boranuide, deuterated sodium borohydride | |
[BH4-].[Na+] |
Specifications
Powder and/or Chunks | |
97.5% min. | |
>300.0°C | |
BH4Na | |
MFCD00003519 | |
sodium borodeuteride, sodium tetradeuterioboron, sodium borohydride-d4, sodium 2h4 tetrahydroborate 1-, sulfatase, borate 1-, tetrahydro-d4-, sodium, nabd4, sodium 2h4 boranuide, deuterated sodium borohydride | |
[BH4-].[Na+] | |
37.83 | |
41.86 | |
NMR |
Gray to White | |
15681-89-7 | |
98 atom% D | |
NaBD4 | |
03,281 | |
YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
sodium boranuide | |
23673181 | |
98 atom% D | |
Sodium borodeuteride, 98 atom % D |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 239-764-7
- Sodium borodeuteride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H261-In contact with water releases flammable gases.
Precautionary Statement
- P223-Do not allow contact with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only