Learn More
Sodium borohydride, (VenPure(TM) solution), 12% aq. solution, Thermo Scientific Chemicals
£16.10 - £125.00
Specifications
Boiling Point | 130.0°C to 135.0°C |
---|---|
Chemical Name or Material | Sodium borohydride |
Name Note | VenPure™ solution, 12% aq. solution |
CAS | 16940-66-2, 1310-73-2, 67-56-1, 7732-18-5 |
Molecular Formula | BH4Na |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13226339
|
Thermo Scientific Acros
448511000 |
100 mL |
£16.10
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13266339
|
Thermo Scientific Acros
448515000 |
500 mL |
£39.30
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
13236339
|
Thermo Scientific Acros
448510025 |
2.5 L |
£125.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
130.0°C to 135.0°C | |
VenPure™ solution, 12% aq. solution | |
BH4Na | |
MFCD00003518 | |
SBH, Sodium tetrahydroborate | |
YOQDYZUWIQVZSF-UHFFFAOYSA-N | |
sodium boranuide | |
37.82 | |
1.4000g/mL |
Sodium borohydride | |
16940-66-2, 1310-73-2, 67-56-1, 7732-18-5 | |
NaBH4 | |
15,8727 | |
Solubility in water: completely soluble | |
[BH4-].[Na+] | |
37.83 | |
1.4 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May damage fertility.
May damage the unborn child.
May be corrosive to metals.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: Rinse mouth.
Do NOT induce
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 241-004-4
RUO – Research Use Only