missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromate, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11377697
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Sodium bromate is used as an oxidant to convert tetrahydropyranyl ethers to carbonyl compounds. It acts as an analytical reagent. It is also used for dissolving gold from its ores, boiler cleaning, oxidation of sulfur and vat dyes. Further, it is used in hair treatment formulations as a hair-permanent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-38-0
Molecular Formula BrNaO3
Molecular Weight (g/mol) 150.891
MDL Number MFCD00003476
InChI Key XUXNAKZDHHEHPC-UHFFFAOYSA-M
Synonym sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french
PubChem CID 23668195
ChEBI CHEBI:75229
IUPAC Name sodium;bromate
SMILES [O-]Br(=O)=O.[Na+]

Specifications

Melting Point 381°C (decomposition)
Color White
Physical Form Crystals
Quantity 100 g
Assay Percent Range 99.5% (metals basis)
Solubility Information Soluble in water (2.5 parts in cold water,1.1 part in boiling water)Soluble in water and ammonia. Insoluble in ethanol,acetone and ether.
Formula Weight 150.9
Odor Odorless
Refractive Index 1.594
Appearance White crystalline
Density 3.34 g/cm3
Chemical Name or Material Sodium bromate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'