missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromate, 99+%, extra pure, Thermo Scientific Chemicals

£21.41 - £219.09

Chemical Identifiers

CAS 7789-38-0
Molecular Formula BrNaO3
Molecular Weight (g/mol) 150.9
MDL Number MFCD00003476
InChI Key XUXNAKZDHHEHPC-UHFFFAOYSA-M
Synonym sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french
PubChem CID 23668195
ChEBI CHEBI:75229
IUPAC Name sodium;bromate
SMILES [O-]Br(=O)=O.[Na+]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10072730
View Documents
Thermo Scientific Acros
208871000
100 g Glass bottle
£21.41
100g
Please to purchase this item. Need a web account? Register with us today!
10082730
View Documents Promotion Details
Thermo Scientific Acros
208875000
500 g Glass bottle
£57.79
500g
Please to purchase this item. Need a web account? Register with us today!
13488458
View Documents
Thermo Scientific Acros
208870025
2.5 kg Glass bottle
£219.09
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

7789-38-0
150.9
XUXNAKZDHHEHPC-UHFFFAOYSA-M
23668195
sodium;bromate
BrNaO3
MFCD00003476
sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french
CHEBI:75229
[O-]Br(=O)=O.[Na+]

Specifications

381.0°C
White
99+%
NaBrO3
01,1055
sodium bromate, bromic acid, sodium salt, dyetone, bromate de sodium, sodium bromate dot, neutralizer k-126, neutralizer k-140, neutralizer k-938, sodium bromate nabro3, bromate de sodium french
XUXNAKZDHHEHPC-UHFFFAOYSA-M
sodium;bromate
23668195
150.9
Extra Pure
3.3300g/mL
7789-38-0
Fine Crystalline Powder
BrNaO3
MFCD00003476
15,8728
Solubility in water: 364g/L (20°C). Other solubilities: insoluble in alcohol , ether and aceton
[O-]Br(=O)=O.[Na+]
150.9
CHEBI:75229
≥99%
3.33
Sodium bromate, 99+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May intensify fire; oxidizer.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse caut

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 232-160-4

missing translation for 'rtecsNumber' : EF8750000

missing translation for 'tsca' : TSCA

Product Identifier
  • Sodium bromate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only