missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bromide, 97%, Thermo Scientific Chemicals
£33.20 - £84.60
Chemical Identifiers
CAS | 7647-15-6 |
---|---|
Molecular Formula | BrNa |
Molecular Weight (g/mol) | 102.89 |
MDL Number | MFCD00003475 |
InChI Key | JHJLBTNAGRQEKS-UHFFFAOYSA-M |
Synonym | sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a |
PubChem CID | 253881 |
ChEBI | CHEBI:63004 |
IUPAC Name | sodium bromide |
SMILES | [Na+].[Br-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11361569
|
Thermo Scientific Alfa Aesar
014037.36 |
500 g |
£33.20
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11371569
|
Thermo Scientific Alfa Aesar
014037.A3 |
2 kg |
£84.60
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sodium bromide is used in the preparation of bromides and as a source of bromide ion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7647-15-6 | |
102.89 | |
JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
253881 | |
sodium bromide |
BrNa | |
MFCD00003475 | |
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
CHEBI:63004 | |
[Na+].[Br-] |
Specifications
1390°C | |
7647-15-6 | |
97% | |
MFCD00003475 | |
Soluble in water,alcohol | |
[Na+].[Br-] | |
102.89 | |
CHEBI:63004 | |
1.6412 | |
3.203 g/mL |
755°C | |
Granules | |
BrNa | |
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
sodium bromide | |
253881 | |
102.89 | |
Hygroscopic | |
Sodium bromide |
Safety and Handling
missing translation for 'einecsNumber' : 231-599-9
missing translation for 'rtecsNumber' : VZ3150000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only