missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium bromide, 99.5%, for analysis, Thermo Scientific Chemicals
£23.92 - £322.61
Chemical Identifiers
CAS | 7647-15-6 |
---|---|
Molecular Formula | BrNa |
Molecular Weight (g/mol) | 102.89 |
MDL Number | MFCD00003475 |
InChI Key | JHJLBTNAGRQEKS-UHFFFAOYSA-M |
Synonym | sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a |
PubChem CID | 253881 |
ChEBI | CHEBI:63004 |
SMILES | [Na+].[Br-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10107903
|
Thermo Scientific Acros
246901000 |
100 g | Plastic bottle |
£23.92
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10174620
|
Thermo Scientific Acros
246905000 |
500 g | Plastic bottle |
£83.31
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10115793
|
Thermo Scientific Acros
246900025 |
2.5 kg | Plastic bottle |
£322.61
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
7647-15-6 | |
102.89 | |
JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
253881 | |
[Na+].[Br-] |
BrNa | |
MFCD00003475 | |
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
CHEBI:63004 |
Specifications
1390.0°C | |
7647-15-6 | |
5 to 8.8 (5% soln. at 20°C) | |
99.5% min. (Argentometry) | |
NaBr | |
12,445; 13,46 | |
sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a | |
JHJLBTNAGRQEKS-UHFFFAOYSA-M | |
sodium;bromide | |
253881 | |
102.89 | |
Analytical |
755.0°C | |
White | |
Crystalline Powder | |
BrNa | |
MFCD00003475 | |
15, 8729 | |
Solubility in water: 905g/L (20°C). Other solubilities: 1160g/L water (50°C), 1210g/L water (100°C), 1g/6 mL methanol, 7g/100 mL 95% ethanol | |
[Na+].[Br-] | |
102.89 | |
CHEBI:63004 | |
99.5% | |
Sodium bromide, p.a. |
Safety and Handling
HYGROSCOPIC
missing translation for 'einecsNumber' : 231-599-9
missing translation for 'rtecsNumber' : VZ3150000
missing translation for 'tsca' : TSCA
RUO – Research Use Only