missing translation for 'onlineSavingsMsg'
Learn More

Sodium bromide, 99+%, extra pure, anhydrous, Thermo Scientific Chemicals

Product Code. 10063573
Click to view available options
Quantity:
100 g
1 kg
5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-15-6
Molecular Formula BrNa
Molecular Weight (g/mol) 102.89
MDL Number MFCD00003475
InChI Key JHJLBTNAGRQEKS-UHFFFAOYSA-M
Synonym sodium bromide, bromide salt of sodium, sodium bromide nabr, sedoneural, sodiumbromide, trisodium tribromide, bromnatrium, nabr, bromnatrium german, caswell no. 750a
PubChem CID 253881
ChEBI CHEBI:63004
IUPAC Name sodium bromide
SMILES [Na+].[Br-]

Specifications

Boiling Point 1390.0°C
Melting Point 755.0°C
Color White
pH 5 to 7 (5% soln. at 20°C)
Physical Form Crystalline Powder
Quantity 5 kg
Assay Percent Range 99% min. (on dry substance) (Argentometry)
Linear Formula NaBr
Fieser 12,445; 13,46
Merck Index 15, 8729
Solubility Information Solubility in water: 905g/L (20°C). Other solubilities: 1160g/L water (50°C),1210g/L water (100°C),1g/6 mL methanol,7g/100 mL 95% ethanol
Formula Weight 102.89
Percent Purity ≥99%
Grade Extra Pure
Loss on Drying 1% max. (120°C)
Packaging Plastic bottle
Chemical Name or Material Sodium bromide, Anhydrous
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.